Order Entry
Belgium
ContactUsLinkComponent
Ethylenediamine
Flammable
Corrosive
Health
Irritant
Ethylenediamine
Catalog # ROTH3049.1
Supplier:  Roth Carl
CAS Number:  
undefined
Flammable
Corrosive
Health
Irritant
Ethylenediamine
Catalog # ROTH3049.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

About this item

 :
 :60.1 g/mol
Merck Index:13,03829
Boiling Pt:117 °C (1013 hPa)
Melting Pt:8.5 °C
Density:0.898 g/cm³ (20 °C)
Flash Pt:34 °C
Storage Temperature:Ambient
EINECS:203-468-6
UN:1604
ADR:8,II
 :GHS02
 :GHS05
 :GHS08
 :GHS07
CAS Number:107-15-3
Formula:C₂H₈N₂
MDL Number:MFCD00008204