Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Taurine
Taurine ≥99%
Taurine ≥99%
:
Thermo Fisher Scientific
Taurine ≥99%
:
Thermo Fisher Scientific
:
107-35-7
:
:
C₂H₇NO₃S
:
MFCD00008197
:
125.15 g/mol
:
14,9074
:
>300 °C
:
203-483-8
:
GHS07
:
Ambient
msds_ALFAA12403
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 40…60 °C, PESTINORM® for capillary GC analysis
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
: Merck
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
: Merck
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
: Merck
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
: VWR Chemicals
Petroleum spirit, 40…60 °C, EMSURE® ACS, ISO for analysis, Supelco®
: Merck
Chloroform 99+% stabilised, extra pure
: Thermo Fisher Scientific
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
1,5-Diaminonaphthalene ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals