
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # AMBI1906
Supplier: Ambion
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
About this item
:
:121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
:GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679