Order Entry
Belgium
ContactUsLinkComponent
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Millipore®
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Millipore®
Catalog # 1.03252.0250
Supplier:  Merck
CAS Number:  
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Millipore®
Catalog # 1.03252.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :209.24 g/mol
Melting Pt:102…103 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:230-237-7
CAS Number:6976-37-0
Formula:C₈H₁₉NO₅
MDL Number:MFCD00002853