Order Entry
Belgium
ContactUsLinkComponent
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99%, Ultrapure
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99%, Ultrapure
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99%, Ultrapure
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • DNase:
    none detected
  • RNase:
    none detected
  • Melting Point:
    102 – 106 °C
  • Moisture (KF):
    ≤1%
  • pH (1.0%, Water) @25 °C:
    8.8 – 9.6
  • pKa @25 °C:
    6.45 – 6.65
  • Protease:
    none
  • Purity (Titration):
    >99%

Specification Test Results

Related Information

Frequently Bought Together