ContactUsLinkComponent
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 98%
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 98%
Certificates
undefined
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 98%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥97.5 %