Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

VWR® Basic ESD Bouffant Caps

White, 580 mm
  113-8900
VWR® Basic ESD Bouffant Caps
  113-8900
VWR® Basic ESD Bouffant Caps
VWR® Basic ESD Bouffant Caps

White, 580 mm

 
106,00 €
   
1 * 1.000 ST
 

These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Anti-static properties

Weight: 10 g/m²

 : 100 pieces per bag, 10 bags per case.

 
  • Ø:
    580 mm
  • Colour:
    White
  • Cat. No.:
    113-8900
  • Anti-static:
    +
  • Garment application:
    General Purpose|Electronics/semi-conductor|Industrial|Food
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.