Orders LinkContactUsLinkComponent
VWR® Basic, Bouffant Caps, Latex-Free
VWR® Basic, Bouffant Caps, Latex-Free
VWR® Basic, Bouffant Caps, Latex-Free
Supplier:  VWR Collection
Certificates
VWR® Basic, Bouffant Caps, Latex-Free
VWR® Basic, Bouffant Caps, Latex-Free
18 Options Available Below
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Product Family Options
Product Information
  • Colour
    Ø
    Availability
    Price
  • Blue
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 70,00 €
    70,00 €
    1 * 1.000 ST
       70,00 €
  • Blue
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 103,00 €
    103,00 €
    1 * 1.000 ST
       103,00 €
  • Blue
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 105,00 €
    105,00 €
    1 * 1.000 ST
       105,00 €
  • Green
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
  • Green
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 103,00 €
    103,00 €
    1 * 1.000 ST
       103,00 €
  • Green
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 105,00 €
    105,00 €
    1 * 1.000 ST
       105,00 €
  • Orange
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
  • Orange
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,30 €
    92,30 €
    1 * 1.000 ST
       92,30 €
  • Orange
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 93,80 €
    93,80 €
    1 * 1.000 ST
       93,80 €
  • Red
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
  • Red
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,30 €
    92,30 €
    1 * 1.000 ST
       92,30 €
  • Red
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 93,80 €
    93,80 €
    1 * 1.000 ST
       93,80 €
  • White
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 70,40 €
    70,40 €
    1 * 1.000 ST
       70,40 €
  • White
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 103,00 €
    103,00 €
    1 * 1.000 ST
       103,00 €
  • White
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,90 €
    80,90 €
    1 * 1.000 ST
       80,90 €
  • Yellow
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
  • Yellow
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,30 €
    92,30 €
    1 * 1.000 ST
       92,30 €
  • Yellow
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 105,00 €
    105,00 €
    1 * 1.000 ST
       105,00 €