Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Trifluoroacetic acid ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™
 
undefined
 
 
Trifluoroacetic acid ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™
 

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Assay:
    99.0 - 101.0 %
  • Non-volatile matter:
    Max. 0.005 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Suitability:
    complying
  • APHA:
    Max. 25

 

 

Frequently Bought Together