Order Entry
Belgium
ContactUsLinkComponent
tri-Potassium citrate monohydrate 99.0-100.5% (by perchloric acid titration), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E332, SAFC®
tri-Potassium citrate monohydrate 99.0-100.5% (by perchloric acid titration), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E332, SAFC®
Certificates
undefined
tri-Potassium citrate monohydrate 99.0-100.5% (by perchloric acid titration), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E332, SAFC®

Specification Test Results

  • Assay (Perchloric Acid Titration, Calculated on Dried Substance):
    99.0 - 100.5%
  • Assay (Perchloric Acid Titration After Drying):
    99.0 - 100.5%
  • Identity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Alkalinity:
    Passes Test
  • Acidity or Alkalinity:
    Passes Test
  • pH-Value (5%; Water):
    7.5 - 9.0
  • Chloride (Cl):
    ≤0.0050%
  • Sulfate (SO₄):
    ≤0.0150%
  • Heavy Metals (As Pb):
    ≤0.0005%
  • Al (Aluminium):
    ≤0.0001%
  • As (Arsenic):
    ≤0.0001%
  • Hg (Mercury):
    ≤0.0001%
  • Na (Sodium):
    ≤0.3%
  • Pb (Lead):
    ≤0.0001%
  • Oxalate (C₂O₄):
    ≤0.009%
  • Tartrate (C₄H₄O₆):
    Passes Test
  • Residual Solvents (CH Q3C):
    Excluded by Manufacturing Process
  • Readily Carbonisable Substance:
    Passes Test
  • Loss On Drying (180 °C):
    4.0 - 6.0%
  • Water:
    4.0 - 7.0%

Specification Test Results

Related Information

Frequently Bought Together