Order Entry
Belgium
ContactUsLinkComponent
di-Potassium oxalate monohydrate, EMSURE® ACS for analysis, Supelco®
Irritant
di-Potassium oxalate monohydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates
Irritant
di-Potassium oxalate monohydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    360 g/l (20 °C) (anhydrous substance)
  • Molar Mass:
    184.24 g/mol
  • Bulk Density:
    700 - 1100 kg/m3
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.13 g/cm3 (20 °C)
  • pH:
    7.0 - 8.5 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together