Potassium oxalate hydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Oxalic acid potassium salt hydrate, Ethanedioic acid potassium salt
CAS Number:6487-48-5
:
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033
Density:2.13
Molecular Weight:166.22 g/mol (anhy)
Melting Pt:>300 °C
ADR:6.1 III
Merck Index:14,7651
EINECS:209-506-8
UN:2811
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together