Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Malonic acid

Glass bottle for solids, 1 kg
Catalog # MOLEM13843683
Irritant
Marketsource
Supplier:   Molekula
Malonic acid
Catalog # MOLEM13843683
Supplier:  Molekula
Marketsource
CAS Number:  141-82-2
Malonic acid

Glass bottle for solids, 1 kg

CAS Number:  141-82-2
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1,3-Propanedioic acid
  • CAS Number:141-82-2
  • Formula:C₃H₄O₄
  • MDL Number:MFCD00002707
  • Molecular Weight:104.06 g/mol
  • Merck Index:13,05732
  • Boiling Pt:387 °C (1013 hPa)
  • Melting Pt:136 °C
  • Density:1.669 g/cm³ (20 °C)
  • Flash Pt:172 °C
  • Storage Temperature:Ambient
  • EINECS:205-503-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.