

Glycolic acid for synthesis, Sigma-Aldrich®
: Merck
:79-14-1
:
:C₂H₄O₃
:MFCD00004312
:76.05 g/mol
:13,04511
:266 °C (1013 hPa)
:79…80 °C
:1.49 g/cm³ (20 °C)
:201-180-5
:3261
:8,II
:M-Clarity™ quality level = MQ200
:>300 °C
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:76 - 80 °C
- Molar Mass:76.05 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:100 °C (decomposition)
- Vapor Pressure:10.8 hPa (80 °C)
- Flash Point:300 °C (decomposition)
- Density:1.49 g/cm3 (25 °C)
- pH:2 (50 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together