

Silver(I) acetate ≥99%
Catalog # STRE93-4701-50G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:166.91 g/mol
Merck Index:12,08648
Melting Pt: Dec.
Density:3.26 g/cm³ (20 °C)
EINECS:209-254-9
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:563-63-3
Formula:C₂H₃AgO₂
MDL Number:MFCD00012446