Order Entry
Belgium
ContactUsLinkComponent
 
 
Ammonium acetate ≥99.99% (trace metals basis)
 
undefined
 
Ammonium acetate ≥99.99% (trace metals basis)

 

  • Appearance of the substance:
    complying
  • Identity (IR):
    complying
  • Assay:
    Min. 97.0 %
  • pH (5 %, 25 °C):
    6.7 - 7.3
  • Sulfated ash:
    Max. 0.01 %
  • Water insoluble matter:
    Max. 0.005 %
  • Metal trace analysis (ICP):
    Max. 100 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Nitrate (NO₃):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.001 %

 

 

Frequently Bought Together