Order Entry
Belgium
ContactUsLinkComponent
 
Sodium formate ≥98%, GPR RECTAPUR®
  27869.297
 :  
undefined
Sodium formate ≥98%, GPR RECTAPUR®
  27869.297
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • NH4 (Ammonium):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 5 ppm

 

 

 :
 :68.01 g/mol
 :13,08694
 :257.31 °C
 :1.919 g/cm³ (20 °C)
 :Ambient
 :205-488-0
 :01-2119486468-21
 :141-53-7
 :HCO₂Na
 :MFCD00013101