Order Entry
Belgium
ContactUsLinkComponent
Sodium formate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Sodium formate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Sodium formate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    550 g/l (20 °C)
  • Melting Point:
    253 °C
  • Molar Mass:
    635 kg/m3
  • Density:
    1.92 g/cm3 (20 °C)
  • pH:
    9 - 10 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together