Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rubidium acetate
Rubidium acetate ≥99% (Rb basis)
Rubidium acetate ≥99% (Rb basis)
Rubidium acetate ≥99% (Rb basis)
STRE93-3701-10G
:
Strem Chemicals, Inc.
:
Rubidium acetate ≥99% (Rb basis)
STRE93-3701-10G
:
Strem Chemicals, Inc.
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
144.51 g/mol
:
246 °C
:
209-255-4
:
563-67-7
:
H₃CCOORb
:
MFCD00013058