Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium acetate

Sodium acetate

Glass bottle for solids, 500 g
Catalog # SERA39571.01
Marketsource
Supplier:   Serva
Sodium acetate
Catalog # SERA39571.01
Supplier:  Serva
Marketsource
CAS Number:  127-09-3
Sodium acetate

Glass bottle for solids, 500 g

CAS Number:  127-09-3
Price
55,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Acetic acid sodium salt
  • CAS Number:127-09-3
  • Formula:H₃CCOONa
  • MDL Number:MFCD00012459
  • Molecular Weight:82.03 g/mol
  • Merck Index:12,08711
  • Boiling Pt:881.4 °C (1013 hPa)
  • Melting Pt:324 °C
  • Density:1.528 g/cm³ (20 °C)
  • Flash Pt:>250 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:204-823-8
  • REACH:01-2119485123-42
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications