Order Entry
Belgium
ContactUsLinkComponent
 
Sodium acetate 99.0-101.0%, EMPROVE® ESSENTIAL USP, FCC, E262, extra pure, SAFC®
 
Sodium acetate 99.0-101.0%, EMPROVE® ESSENTIAL USP, FCC, E262, extra pure, SAFC®
 

 

  • Assay (perchloric acid titration, calculated on dried substance):
    99.0 - 101.0 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • In water insoluble matter:
    ≤ 0.05 %
  • Alkalinity:
    ≤ 0.2 %
  • pH-value (1 %, water):
    8.0 - 9.5
  • pH-value (3 %, water):
    7.5 - 9.2
  • Chloride (Cl):
    ≤ 0.01 %
  • Sulphate (SO₄):
    ≤ 0.0050 %
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • As (Arsenic):
    ≤ 0.0002 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • K (Potassium):
    passes test
  • Pb (Lead):
    ≤ 0.0002 %
  • Ca + Mg (calcium and magnesium):
    passes test
  • free acetic acid:
    ≤ 0.4 %
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • formic acid, formate and other oxidizable impurities:
    ≤ 0.1000 %
  • Loss on drying (120 °C):
    ≤ 1.0 %

 

Frequently Bought Together