Order Entry
Belgium
ContactUsLinkComponent
Sodium acetate, anhydrous, Suprapur® for trace analysis, Supelco®
Sodium acetate, anhydrous, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
Sodium acetate, anhydrous, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    365 g/l (20 °C)
  • Melting Point:
    324 °C (decomposition)
  • Molar Mass:
    82.04 g/mol
  • Boiling Point:
    400 °C (decomposition)
  • Vapor Pressure:
    (20 °C)
  • Flash Point:
    250 °C
  • Density:
    1.52 g/cm3 (20 °C)
  • pH:
    7.5 - 9.0 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    607 °C

Specification Test Results

Related Information

Frequently Bought Together