ContactUsLinkComponent
Cell extract HL60 (non-induced)
  ENZOBMLSW1010200
 :  
Cell extract HL60 (non-induced)
  ENZOBMLSW1010200
 :  
  • Pk:
    200 µl
  • Description:
    Immunoblotting standard, HL60 cell extract (non-induced)
  • Seq:
    02
Whole-cell extract of human HL-60 leukemia cells.
  • Band with MW=116 kDa, which represents intact PARP, is detected by Western blotting
  • Ready to use for SDS-PAGE
  • Use approximately 20 µl sample per lane (~75,000 cells)