Order Entry
Belgium
ContactUsLinkComponent
Isopropyl myristate for synthesis, Sigma-Aldrich®
Isopropyl myristate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Isopropyl myristate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
     - 0.05 mg/l (20 °C)
  • Molar Mass:
    270.45 g/mol
  • Boiling Point:
    140 °C (3 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    150 °C
  • Refractive Index:
    1.434 (20 °C, 589 nm)
  • Density:
    0.85 g/cm3 (20 °C)
  • Ignition Point:
    300 °C
  • Viscosity kinematic:
    6.57 mm2/s (20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together