Isopropyl myristate Ph. Eur., NF, SAFC®
Catalog # 8.17066.2500
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:270.46 g/mol
Merck Index:13,05235
Boiling Pt:320 °C (1013 hPa)
Melting Pt:0…1 °C
Density:0.852 g/cm³ (20 °C)
Flash Pt:150 °C
Storage Temperature:Refrigerator
EINECS:203-751-4
CAS Number:110-27-0
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008982