Methyl hexanoate ≥99%
Purity: 99% CAS No.: 106-70-7 Molecular Formula: CH3(CH2)4CO2CH3 Formula Weight: 130.19 Flash Point: 45°(113°F) Refractive Index: 1.4050 Melting Point: -71° Boiling Point: 150-151° Beilstein Reference: 1744683 UN No.: 3272 MDL No.: MFCD00009510
: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
- :106-70-7
- :C₇H₁₄O₂
- :MFCD00009510
- :130.19 g/mol
- :150…151 °C
- :–71 °C
- :0.885
- :45 °C (113 °F)
- :Ambient
- :203-425-1
- :3272
- :3,III
Purity: 99% CAS No.: 106-70-7 Molecular Formula: CH3(CH2)4CO2CH3 Formula Weight: 130.19 Flash Point: 45°(113°F) Refractive Index: 1.4050 Melting Point: -71° Boiling Point: 150-151° Beilstein Reference: 1744683 UN No.: 3272 MDL No.: MFCD00009510
: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
- :106-70-7
- :C₇H₁₄O₂
- :MFCD00009510
- :130.19 g/mol
- :150…151 °C
- :–71 °C
- :0.885
- :45 °C (113 °F)
- :Ambient
- :203-425-1
- :3272
- :3,III