Order Entry
Belgium
ContactUsLinkComponent
Hydrogen peroxide colorimetric detection kit
Hydrogen peroxide colorimetric detection kit
Catalog # ENZOADI907015
Supplier:  Enzo Life Sciences
CAS Number:  
Hydrogen peroxide colorimetric detection kit
Catalog # ENZOADI907015
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Hydrogen peroxide colorimetric detection kit
  • Seq:
    02
  • Sensitivity:
    51,25 ng/ml (range 106,25 - 3400 ng/ml)

Specifications

About this item

Hydrogen peroxide colorimetric detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide in a wide variety of samples.