ContactUsLinkComponent
Clamp for analogue and digital shakers
Clamp for analogue and digital shakers
Catalog # OHAU30400104
Supplier:  OHAUS
undefined
Clamp for analogue and digital shakers
Catalog # OHAU30400104
Supplier:  OHAUS
Specifications
  • For:
    Analogue/digital 16 kg, 23 kg, 45 kg and 68 kg shakers
  • Description:
    Microplate clamp, stainless steel 304
  • Pk:
    1
Specifications
About this item
Microplate clamp attaches to the platform allowing micrplates to be held on the platform.