Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Ethyl acetate ≥99.8%, PESTINORM® for capillary GC analysis
  83963.320
   Safer Human and Environmental Health
 :  
undefined
 
 
Ethyl acetate ≥99.8%, PESTINORM® for capillary GC analysis
  83963.320
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue (100°C):
    Max. 0.0005 %
  • Water:
    Max. 0.05 %
  • Organic residue (as Octanol) (GC/FID):
    Max. 10 ng/l
  • Halogenated residue (as Lindane)(GC/ECD):
    Max. 5 ng/l

 

 

 :
 :88.11 g/mol
 :13,03792
 :77.1 °C (1013 hPa)
 :–83 °C
 :0.902 g/cm³ (20 °C)
 :–4 °C
 :Ambient
 :205-500-4
 :1173
 :3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :01-2119475103-46
 :GHS02
 :GHS07
 :141-78-6
 :CH₃COOC₂H₅
 :MFCD00009171