

Butyl acetate ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:123-86-4
:
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445
:116.16 g/mol
Merck Index:13,01534
Boiling Pt:126.2 °C (1.013 hPa)
Melting Pt:–78 °C
Density:0.881 g/cm³ (20 °C)
Flash Pt:10.4 %(V)
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.65 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 320 nm:Max. 0.01
- Absorbance at 300 nm:Max. 0.02
- Absorbance at 275 nm:Max. 0.04
- Absorbance at 260 nm:Max. 0.20
- Absobance at 254 nm:Max. 1.0
- Explosion Limit:7.6 %
- Autoignition Temperature:790 °F
- Vapor Pressure:15 mmHg ( 25 °C)
Specification Test Results
Frequently Bought Together