Urea
Marketsource
Supplier: Serva
About this item
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
3 Options Available Below
Product Details & Documents
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.SERA24524.01Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.SERA24524.02Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.SERA24524.03Marketsource



