Order Entry
Belgium
ContactUsLinkComponent
Urea for synthesis, Sigma-Aldrich®
Urea for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Urea for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    590 g/l (20 °C)
  • Melting Point:
    133 °C
  • Molar Mass:
    60.06 g/mol
  • Bulk Density:
    720 - 760 kg/m3
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Density:
    1.34 g/cm3 (20 °C)
  • pH:
    9 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together