Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Urea

Marketsource
Supplier:   Ambion
Urea
Supplier:  Ambion
Marketsource
Certificates
Urea

2 Options Available Below

Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource