Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
 
undefined
 
 
 
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™

 

  • Suitability for the GC Headspace:
    complying
  • Assay (GC):
    Min. 99.9 %
  • Refractive index (n 20/D):
    1.429 - 1.431
  • Water (Karl Fischer):
    Max. 0.03 %
  • Absorbance at 270 nm:
    Max. 0.700
  • Absorbance at 275 nm:
    Max. 0.250
  • Absorbance at 300 nm:
    Max. 0.070
  • Absorbance at 320 nm:
    Max. 0.022

 

 

Frequently Bought Together