Orders LinkContactUsLinkComponent
MULTIVIEW® Microarray Slides
MULTIVIEW® Microarray Slides
Supplier:   Enzo Life Sciences
MULTIVIEW® Microarray Slides
Supplier:  Enzo Life Sciences
Certificates
MULTIVIEW® Microarray Slides
2 Options Available Below
Product Details & Documents
These microarray slides with eleven human tissue specimens includes placenta, liver, tonsil, colon, skin, brain, breast (fibroadenoma), prostate, thyroid, kidney and fallopian tube.
  • High-consistency specimen and slide preparation for reduced maintenance
  • Fixed in formalin and paraffin embedded
  • Sections are placed on a SuperFrost Plus slide
  • Cores are approximately 2 mm in diameter
  • Tissue sections are 3 µm thick
Suitable for use with common FFPE tissue protocols as rapid screening tools for the optimisation or validation of stains in a broad-spectrum of immunohistochemical procedures.
Product Family Options
Product Information
  • Availability
    Price
  • ENZOADI9500200025
    Universal tissue microarray slides, 25 slides
    Universal tissue microarray slides, 25 slides
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 896,00 €
    896,00 €
    1 * 5 ST
       896,00 €
  • ENZOADI9500200050
    Universal tissue microarray slides, 50 slides
    Universal tissue microarray slides, 50 slides
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 560,00 €
    1 560,00 €
    1 * 5 ST
       1 560,00 €