Order Entry
Belgium
ContactUsLinkComponent
Sodium pyruvate ≥99%, Reagent Grade
Sodium pyruvate ≥99%, Reagent Grade
Supplier:  VWR Chemicals
Certificates
undefined
Sodium pyruvate ≥99%, Reagent Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Free Acid:
    ≤1
  • Heavy Metals (as Pb):
    ≤0.01%
  • Water Content:
    <1%
  • Purity:
    ≥99%

Specification Test Results

Related Information

Frequently Bought Together