Order Entry
Belgium
ContactUsLinkComponent
 
Sodium pyruvate 99+%
 
undefined
Sodium pyruvate 99+%

 

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 %
  • Heavy metals:
    ≤10 ppm
  • Solubility:
    (10 % in water) Clear colorless to light yellow
  • Titration with HClO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤20 ppm
  • Sulfate (SO4):
    ≤20 ppm
  • Arsenic (As):
    ≤1 ppm

 

 

Frequently Bought Together