Acetone ≥99.8% (by GC), CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:55…57 °C (1.013 hPa)
Melting Pt:–95 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:14.3 %(V)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Refractive Index:n20/D 1.359 (lit.)
- Vapor Density:2 (vs air)
- Assay (GC):Min. 99.8 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.2 %
- Suitability for GC-MS of dioxines, furanes, PCPs:complying
Specification Test Results
Frequently Bought Together