

Acetone ≥99.9%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
EINECS:200-662-2
UN:1090
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Acidity:Max. 0.0005 meq/g
- Evaporation residue (100°C):Max. 0.0005 %
- Organic residue (as Octanol) (GC/FID):Max. 10 ng/ml
- Halogenated residue (as Lindane)(GC/ECD):Max. 5 ng/l
- Water:Max. 0.3 %
Specification Test Results
Frequently Bought Together