Order Entry
Belgium
ContactUsLinkComponent
 
Sebaconitrile for synthesis, Sigma-Aldrich®
  8.14070.0025
 :  Merck
 :  
Sebaconitrile for synthesis, Sigma-Aldrich®
  8.14070.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    7 - 8 °C
  • Molar Mass:
    164.24 g/mol
  • Boiling Point:
    199 - 204 °C (20 hPa)
  • Flash Point:
    93 °C
  • Refractive Index:
    1.4474 (20 °C, 589 nm)
  • Density:
    0.91 g/cm3 (20 °C)
  • Synonyms:
    Decanedinitrile, Decanedioic acid dinitrile, Sebaconitrile
  • Hill Formula:
    C10H16N2
  • Chemical Formula:
    CN(CH2)8CN

 

 

 :
 :164.25 g/mol
 :332 °C (1013 hPa)
 :0.91 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :217-494-0
 :1871-96-1
 :C₁₀H₁₆N₂
 :MFCD00019900