Order Entry
Belgium
Orders LinkContactUsLinkComponent
Chloroform

Chloroform

Toxic
Health
Marketsource
Supplier:   Roth Carl
Chloroform
Supplier:  Roth Carl
Marketsource
Certificates
Chloroform

4 Options Available Below

Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-663-8
  • UN:1888
  • ADR:6.1,III
  • REACH:05-2114614906-45
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 33,50 €
    33,50 €
    1 * 1 L
       33,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 66,50 €
    66,50 €
    1 * 2,5 L
       66,50 €

    Specifications:
    Marketsource
    Marketsource
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 239,00 €
    239,00 €
    1 * 10 L
       239,00 €

    Specifications:
    Marketsource
    Marketsource
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 485,00 €
    485,00 €
    1 * 25 L
       485,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.