Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Chloroform

Glass bottle, 2,5 L
Catalog # ROTH7331.1
Toxic
Health
Marketsource
Supplier:   Roth Carl
Chloroform
Catalog # ROTH7331.1
Supplier:  Roth Carl
Marketsource
CAS Number:  67-66-3
Chloroform
Chloroform

Glass bottle, 2,5 L

CAS Number:  67-66-3
Price
Inquire For Price
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-663-8
  • UN:1888
  • ADR:6.1,III
  • REACH:05-2114614906-45
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.