Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

1,2-Dichloroethane

Flammable
Health
Irritant
Marketsource
Supplier:   Roth Carl
1,2-Dichloroethane
Supplier:  Roth Carl
Marketsource
Certificates
1,2-Dichloroethane
1,2-Dichloroethane

3 Options Available Below

Product Details & Documents
Synonyms:  Ethylene chloride, Ethylene dichloride, Dichloroethane
  • CAS Number:107-06-2
  • Formula:ClH₂CCH₂Cl
  • MDL Number:MFCD00000963
  • Molecular Weight:98.96 g/mol
  • Merck Index:13,03831
  • Boiling Pt:83.5 °C (1013 hPa)
  • Melting Pt:–35.5 °C
  • Density:1.254 g/cm³ (20 °C)
  • Flash Pt:13 °C
  • Storage Temperature:Ambient
  • EINECS:203-458-1
  • UN:1184
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
    Marketsource
    Marketsource
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L

    Specifications:
    Marketsource
    Marketsource