Order Entry
Belgium
ContactUsLinkComponent
 
Potassium sorbate 99.0-101.0%, granules, EMPROVE® ESSENTIAL FCC, BP, E202, Ph. Eur., NF suitable for use as excipient, SAFC®
Catalog # 1.05119.9025
CAS Number:  
undefined
Potassium sorbate 99.0-101.0%, granules, EMPROVE® ESSENTIAL FCC, BP, E202, Ph. Eur., NF suitable for use as excipient, SAFC®
Catalog # 1.05119.9025
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (acidimetric; calculated on dried substance):
    99.0 - 101.0 %
  • Identity:
    passes test
  • Appearance of solution (5 %; water):
    passes test
  • Acidity or alkalinity:
    passes test
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • As (Arsenic):
    ≤ 0.0003 %
  • Cd (Cadmium):
    ≤ 0.001 %
  • Cu (Copper):
    ≤ 0.001 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • Pb (Lead):
    ≤ 0.0002 %
  • Zn (Zinc):
    ≤ 0.001 %
  • Aldehydes (as Acetaldehyd):
    ≤ 0.15 %
  • Residual solvents (Ph.Eur./USP/ICH) class 3:
    ≤ 0.5 %
  • Other residual solvents (Ph.Eur./USP/ICH):
    excluded by production process
  • Thermal stability (105 °C):
    passes test
  • Loss on Drying (105°C):
    ≤ 1.0 %

Specification Test Results

About this item

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
 :
 :150.22 g/mol
Merck Index:13,07756
Melting Pt:270 °C
Density:1.36 g/cm³ (20 °C)
EINECS:246-376-1
 :GHS07
Storage Temperature:Ambient
CAS Number:24634-61-5
 :590-00-1
Formula:CH₃CH=CHCH=CHCOOK
MDL Number:MFCD00016546