Potassium sorbate 99.0-101.0% (by acidimetry), EMPROVE® ESSENTIAL FCC, BP, E202, Ph. Eur., NF suitable for use as excipient, SAFC®
Catalog # 1.05118.9025
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Cardboard carton
- Pk:25 kg
- Seq:4283
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (Acidimetric; Calculated on Dried Substance):99.0 - 101.0%
- Identity:Passes Test
- Appearance of Solution (5%; Water):Passes Test
- Acidity or Alkalinity:Passes Test
- Heavy Metals (as Pb):≤0.0010%
- As (Arsenic):≤3 ppm
- Cd (Cadmium):≤10 ppm
- Cu (Copper):≤10 ppm
- Hg (Mercury):≤1 ppm
- Pb (Lead):≤2 ppm
- Zn (Zinc):≤10 ppm
- Aldehydes (as Acetaldehyde):≤0.15%
- Residual Solvents (Ph Eur/USP/ICH) class 3:<0.5%
- Other Residual Solvents (Ph. Eur./USP/ICH):Excluded by Production Process
- Thermal Stability (105 °C):Passes Test
- Loss on Drying (105 °C):≤1.0%
Specification Test Results
About this item
Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
:
:150.22 g/mol
Merck Index:13,07756
Melting Pt:270 °C
Density:1.36 g/cm³ (20 °C)
EINECS:246-376-1
:GHS07
Storage Temperature:Ambient
CAS Number:24634-61-5
:590-00-1
Formula:CH₃CH=CHCH=CHCOOK
MDL Number:MFCD00016546