Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Biotin

D-(+)-Biotin

Glass bottle for solids, 25 mg
Catalog # PIER29130
D-(+)-Biotin
Catalog # PIER29130
CAS Number:  58-85-5
D-(+)-Biotin
D-(+)-Biotin

Glass bottle for solids, 25 mg

CAS Number:  58-85-5
Price
260,00 €
Log in for availability
1 * 25 mg
Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:13,01231
  • Melting Pt:232…233 °C
  • Density:1.41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.