Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Biotin

D-(+)-Biotin

Glass bottle for solids, 1 g
Catalog # A0969.0001
Supplier:   PanReac AppliChem
D-(+)-Biotin
Catalog # A0969.0001
Supplier:  PanReac AppliChem
CAS Number:  58-85-5
D-(+)-Biotin

Glass bottle for solids, 1 g

CAS Number:  58-85-5
Price
89,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:13,01231
  • Melting Pt:232…233 °C
  • Density:1.41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications