Order Entry
Belgium
ContactUsLinkComponent
L(+)-Histidine 99.0-101.0% (by perchloric acid titration), EMPROVE® EXPERT Ph. Eur., USP, JP, SAFC®
L(+)-Histidine 99.0-101.0% (by perchloric acid titration), EMPROVE® EXPERT Ph. Eur., USP, JP, SAFC®
Certificates
undefined
L(+)-Histidine 99.0-101.0% (by perchloric acid titration), EMPROVE® EXPERT Ph. Eur., USP, JP, SAFC®
Certificates

Specification Test Results

  • Assay (acidimetric, calculated on dried substance):
    98.5 - 101.0 %
  • Assay (perchloric acid titration, calculated on dried substance):
    99.0 - 101.0 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    white to almost white fine-crystalline powder or colorless crystals
  • Appearance of solution (50 g/l; water):
    clear and not more intense in color than reference solution BY₇.
  • Appearance of solution (20 g/l, water):
    clear and colorless
  • Spec. rotation (α 20/D, 110 g/l, hydrochloric acid 120 g/l, calc. on dried substance):
    +11.4 to +12.4
  • Spec. rotation (α 25/D, 110 g/l, hydrochloric acid 6 mol/l, calc. on dried substance):
    +12.6 to +14.0
  • Spec. rotation (α 20/D, 110 g/l, hydrochloric acid 6 mol/l, calc. on dried substance):
    +11,8° to +12,8°
  • pH (20 g/l, CO₂-free water):
    7.0 - 8.5
  • Chloride (Cl):
    ≤ 200 ppm
  • Sulfate (SO₄):
    ≤ 200 ppm
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • Cu(Copper):
    ≤ 25 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • Fe (Iron) (ICP):
    ≤ 10 ppm
  • Mn (Manganese):
    ≤ 25 ppm
  • Zn (Zinc):
    ≤ 130 ppm
  • Related Compounds, total (TLC):
    ≤ 0.5 %
  • Related substances (TLC):
    passes test
  • Ninhydrin-positive substances (LC) (any ninhydrin-positive impurity):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC) (ammonium (570 nm)):
    ≤ 0.02 %
  • Ninhydrin-positive substances (LC) (total impurities):
    ≤ 0.5 %
  • Residual solvents (ICH (Q3C)):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (105 °C; 3 h):
    ≤ 0.2 %
  • Bacterial endotoxins:
    < 2.0
  • Microbiological test (total aerobic microbial count (TAMC)):
    ≤ 10²
  • Microbiological test (total yeast and mould count (TYMC)):
    ≤ 10²

Specification Test Results

Related Information

Frequently Bought Together