L(+)-Valine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:72-18-4
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064220
:117.15 g/mol
Merck Index:13,09458
Boiling Pt:217 °C (1013 hPa)
Melting Pt:86 °C
Density:1.284 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-773-6
Specification Test Results
- Solubility:85 g/l (20 °C)
- Melting Point:315 °C
- Molar Mass:117.15 g/mol
- Bulk Density:400 kg/m3
- Density:1.23 g/cm3 (20 °C)
- pH:5.5 - 6.5 (100 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together