ContactUsLinkComponent
L(+)-Valine, Millipore®
L(+)-Valine, Millipore®
Supplier:  Merck
Certificates
L(+)-Valine, Millipore®
Supplier:  Merck
Specification Test Results
  • Ammonium (NH4):
    0.01% max.
  • Foreign Amino Acids:
    0.3% max.
  • Heavy Metals (as Pb):
    0.001% max.
  • Melting Point:
    315°C